Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isosorbide dinitrate | 87-33-2 | MFCD00868238 | 99.9% | 236.14 | C6H8N2O8 | 10 MG
SDP

Catalog No. 5000296134
Encompass_Preferred

Isosorbide dinitrate is a nitric oxide (NO) donor used as a research reagent and analytical standard for studies of vasodilation, myocardial infarction, and cardiac remodeling. It is supplied as a high-purity solid suitable for in vitro and analytical applications.

  • High purity (99.9%).
  • Chemical formula C6H8N2O8; molecular weight 236.14.
  • CAS number 87-33-2.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Store protected from light at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Available as solid packs (5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg) and as 10 mM solutions in DMSO.

Catalog No. 50-002-96134 Supplier Medchemexpress LLC Supplier No. HYB140910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isosorbide dinitrate is a nitric oxide (NO) donor used as a research reagent and analytical standard for studies of vasodilation, myocardial infarction, and cardiac remodeling. It is supplied as a high-purity solid suitable for in vitro and analytical applications.

  • High purity (99.9%).
  • Chemical formula C6H8N2O8; molecular weight 236.14.
  • CAS number 87-33-2.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Store protected from light at 4°C; in solvent, -80°C up to 6 months or -20°C up to 1 month.
  • Available as solid packs (5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg) and as 10 mM solutions in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.