Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isosorbide dinitrate (ISDN) | 87-33-2 | 99.9% | 50 MG
SDP

Catalog No. 5000413237
Encompass_Preferred

Isosorbide dinitrate (ISDN) is an NO donor that prevents LV remodeling and degradation of cardiac function following myocardial infarction (MI). It acts as an NO donor, preventing LV remodeling and degradation of cardiac function post-MI.

  • Molecular weight: 236.14
  • Formula: C6H8N2O8
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 100 mg/mL (423.48 mM)

Catalog No. 50-004-13237 Supplier Medchemexpress LLC Supplier No. HYB140950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isosorbide dinitrate (ISDN) is an NO donor that prevents LV remodeling and degradation of cardiac function following myocardial infarction (MI). It acts as an NO donor, preventing LV remodeling and degradation of cardiac function post-MI.

  • Molecular weight: 236.14
  • Formula: C6H8N2O8
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 100 mg/mL (423.48 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.