Learn More
Medchemexpress LLC Isosorbide mononitrate | 16051-77-7 | 99.9% | 500 MG
SDP
Catalog No. 5000408487
Encompass_Preferred
Isosorbide mononitrate is an orally active nitric acid compound primarily used for angina pectoris. It functions by dilating blood vessels and lowering blood pressure, making it promising for research in heart failure and coronary heart disease.
  • Orally active nitric acid compound
  • Dilates blood vessels
  • Lowers blood pressure
  • Increases viability and proliferation of HUVECs by decreasing apoptosis
  • Elevates expressions of vedf, kdrl, and pdgfr in zebrafish embryos
  • Promising for research in heart failure and coronary heart disease
Catalog No. 50-004-08487 Supplier Medchemexpress LLC Supplier No. HYB0642500MG
Isosorbide mononitrate is an orally active nitric acid compound primarily used for angina pectoris. It functions by dilating blood vessels and lowering blood pressure, making it promising for research in heart failure and coronary heart disease.
  • Orally active nitric acid compound
  • Dilates blood vessels
  • Lowers blood pressure
  • Increases viability and proliferation of HUVECs by decreasing apoptosis
  • Elevates expressions of vedf, kdrl, and pdgfr in zebrafish embryos
  • Promising for research in heart failure and coronary heart disease