Learn More
Medchemexpress LLC Isosulfan blue 50mg | 68238-36-8 | 50MG
SDP
Catalog No. 5000240138
Encompass_Preferred
Isosulfan blue is a blue dye for the identification of lymph vessels during lymphangiography Isosulfan blueis is used during sentinel lymph node biopsies in breast cancer Isosulfan blue is possible to have an allergic reaction during breast cancer operations[1 [2
Catalog No. 50-002-40138 Supplier Medchemexpress LLC Supplier No. HY10796750MG
Isosulfan blue is a blue dye for the identification of lymph vessels during lymphangiography Isosulfan blueis is used during sentinel lymph node biopsies in breast cancer Isosulfan blue is possible to have an allergic reaction during breast cancer operations[1 [2