Learn More
Medchemexpress LLC Isosulfan blue | 68238-36-8 | MFCD00867653 | 99.8% | 566.66 g·mol⁻¹ | C27H31N2NaO6S2 | 100 MG
SDP
Catalog No. 5000242335
Encompass_Preferred
Isosulfan blue is a synthetic lymphatic dye used to delineate lymphatic vessels and identify sentinel lymph nodes during lymphangiography and surgical mapping. Supplied as a high-purity reagent for research and diagnostic workflows, it is provided as a powder or in solution formats for analytical methods and intraoperative visualization.
  • High purity (99.8%) suitable for analytical and diagnostic use.
  • Available as solid powder and as preprepared solution formats.
  • Distinctive blue color for clear lymphatic visualization during procedures.
  • Compatible with common solvents, including DMSO, for solution preparation.
  • Useful as a reference standard for method development and quantification.
  • Provides established chemical identifiers for traceability and reporting.
Catalog No. 50-002-42335 Supplier Medchemexpress LLC Supplier No. HY107967100MG
Isosulfan blue is a synthetic lymphatic dye used to delineate lymphatic vessels and identify sentinel lymph nodes during lymphangiography and surgical mapping. Supplied as a high-purity reagent for research and diagnostic workflows, it is provided as a powder or in solution formats for analytical methods and intraoperative visualization.
  • High purity (99.8%) suitable for analytical and diagnostic use.
  • Available as solid powder and as preprepared solution formats.
  • Distinctive blue color for clear lymphatic visualization during procedures.
  • Compatible with common solvents, including DMSO, for solution preparation.
  • Useful as a reference standard for method development and quantification.
  • Provides established chemical identifiers for traceability and reporting.