Learn More
Cayman Chemical Isotachysterol 3 10mg
SDP
Catalog No. 502910826
Encompass_Preferred
An impurity in the preparation of vitamin D3; stimulates intestinal calcium transport and bone calcium mobilization in control, as well as in anephric, rats fed a low-calcium and vitamin D-deficient diet at 5 UG/animal
Catalog No. 50-291-0826 Supplier Cayman Chemical Supplier No. 3575110 MG
An impurity in the preparation of vitamin D3; stimulates intestinal calcium transport and bone calcium mobilization in control, as well as in anephric, rats fed a low-calcium and vitamin D-deficient diet at 5 UG/animal