Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isotanshinone IIA | 20958-15-0 | 1 MG
SDP

Catalog No. 5000417997
Encompass_Preferred

Isotanshinone IIA is an abietane-type diterpene metabolite. It functions as a non-competitive inhibitor of Protein Tyrosine Phosphatase 1B (PTP1B) activity with an IC50 of 11.4 μM. This compound appears as an orange to orange-red solid and is intended for research use only.

  • High purity: 98.0%
  • IC50: 11.4 μM for PTP1B
  • Molecular weight: 294.34
  • Chemical formula: C19H18O3
  • Appearance: Solid
  • Color: Orange to orange-red
  • Structure classification: Quinones, Naphthalene Quinones
  • Initial source: Plants Leguminosae Samanea saman (Jacq.) Merr., Plants Labiatae
  • Solubility in DMSO: 3 mg/mL (10.19 mM)

Catalog No. 50-004-17997 Supplier Medchemexpress LLC Supplier No. HYN66501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Isotanshinone IIA is an abietane-type diterpene metabolite. It functions as a non-competitive inhibitor of Protein Tyrosine Phosphatase 1B (PTP1B) activity with an IC50 of 11.4 μM. This compound appears as an orange to orange-red solid and is intended for research use only.

  • High purity: 98.0%
  • IC50: 11.4 μM for PTP1B
  • Molecular weight: 294.34
  • Chemical formula: C19H18O3
  • Appearance: Solid
  • Color: Orange to orange-red
  • Structure classification: Quinones, Naphthalene Quinones
  • Initial source: Plants Leguminosae Samanea saman (Jacq.) Merr., Plants Labiatae
  • Solubility in DMSO: 3 mg/mL (10.19 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.