Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isotanshinone IIA | 20958-15-0 | 5 MG
SDP

Catalog No. 5000418469
Encompass_Preferred

Isotanshinone IIA is an abietane-type diterpene metabolite that non-competitively inhibits Protein Tyrosine Phosphatase 1B (PTP1B) activity with an IC50 of 11.4 μM. Intended for research use only, this product is an orange to orange-red solid with 98.0% purity. It has a molecular weight of 294.34 and a chemical formula of C19H18O3.

  • Non-competitive inhibitor of Protein Tyrosine Phosphatase 1B (PTP1B) activity
  • IC50 of 11.4 μM
  • 98.0% purity
  • Appears as an orange to orange-red solid
  • Molecular weight of 294.34
  • Chemical formula of C19H18O3
  • Classified under Quinones and Naphthalene Quinones
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 3 mg/mL (10.19 mM)

Catalog No. 50-004-18469 Supplier Medchemexpress LLC Supplier No. HYN66505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isotanshinone IIA is an abietane-type diterpene metabolite that non-competitively inhibits Protein Tyrosine Phosphatase 1B (PTP1B) activity with an IC50 of 11.4 μM. Intended for research use only, this product is an orange to orange-red solid with 98.0% purity. It has a molecular weight of 294.34 and a chemical formula of C19H18O3.

  • Non-competitive inhibitor of Protein Tyrosine Phosphatase 1B (PTP1B) activity
  • IC50 of 11.4 μM
  • 98.0% purity
  • Appears as an orange to orange-red solid
  • Molecular weight of 294.34
  • Chemical formula of C19H18O3
  • Classified under Quinones and Naphthalene Quinones
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 3 mg/mL (10.19 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.