Learn More
SAI Infusion Technologies Pre-filled Syringes - 1cc Syringe filled with 1ml Saline
SDP
Catalog No. NC2586911
Encompass
Pre-filled Syringes - 1cc Syringe filled with 1ml Saline, white button cap. Sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. These 1cc syringes are pre-filled with 1ml of 0.9% Sodium Chloride, 100/case. Product is for laboratory animal research only.
Catalog No. NC2586911 Supplier SAI Infusion Technologies Supplier No. SC11.0
Pre-filled Syringes - 1cc Syringe filled with 1ml Saline, white button cap. Sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. These 1cc syringes are pre-filled with 1ml of 0.9% Sodium Chloride, 100/case. Product is for laboratory animal research only.