Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ 400 Refrigerated Incubator, 381 L, Stainless Steel

Catalog No. 151031569PM Shop All Fisherbrand Products
Change view
Click to view available options
Model:
Isotemp 400
Capacity (Metric):
381 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Capacity (Metric)
15-103-1569PM Isotemp 400 381 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 15-103-1569PM

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Temperature Range: 10°C to 60°C

These Isotemp units use a Peltier device which can both heat and cool precisely to the desired temperature.
Benefits of Peltier technology:
  • Low energy consumption, especially in temperature range 15 to 25 ºC
  • No hazardous refrigerants
  • No bulky compressor with random vibrations
  • Excellent temperature stability and uniformity

Key Features of Isotemp Peltier Low Temperature Incubators:
  • Access port with silicone plug for independent data logging or for use of electrical equipment inside the unit
  • Peltier device provides precise temperature within chamber – from 10° to 60°C
  • Large easy-to-view fluorescent display with intuitive microprocessor controller
  • Internal glass door for undisturbed sample viewing
  • Readily stackable same-size units without the need for tools or kits
  • Flexible shelving system with different positions to optimize chamber space
  • Lockable casters for easy relocation (large unit)
  • Outstanding temperature uniformity and stability to keep samples at desired condition
  • Wide temperature range: 10° to 60°C
  • Intuitive and easy-to-use microprocessor controller
  • Automatic over temperature alarm – no operator settings required
  • Heated door prevents condensation at lower temperatures
  • Timer function for pre-set power-up or shut off (up to 99 hours)
  • Corrosion resistant stainless steel* interior
  • Easy-to-clean rounded interior edges, designed to reduce contamination risks
  • Flexible shelving with no tools required to set up, adjust or remove
  • Small external dimensions per chamber size reduce required bench / floor space
  • Standard RS232 interface for datalogging (no software provided)
  • 2 year parts and labor warranty
  • cCSAus certified
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 13.5 cu. ft.
Capacity (Metric) 381 L
No. of Shelves 2 std./17 max.
Plug Type Nema 5-15 US
Temperature Range (Metric) 10°C to 60°C
Model Isotemp 400
Temperature Stability ≤ ±0.1 at 20°C
Temperature Uniformity ≤ ±0.4 at 20°C
Dimensions (D x W x H) Interior 524 x 544 x 1335 mm (20.6 x 21.4 x 52.6 in.)
Dimensions (L x W x H) 770** x 778 x 1545** mm (30.3** x 30.6 x 60.8** in.)
** Depth of handle / display not included in depth (65 mm / 2.6 in.); casters not included in height (180 mm / 7.1 in.) – required distance to rear wall: 150 mm / 5.9 in.
Weight (English) 357 lb.
Weight (Metric) 162 kg
Electrical Requirements 120 V, 60 Hz, 480 w; 3.8 A
Wattage 90 w at 20°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.