Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators, Sliding Glass Door

Catalog No. FBG45CPLA Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
45 cu. ft.
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color
FBG45CPLA 45 cu. ft. White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBG45CPLA

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ Chromatography Refrigerators for General Purpose Use are designed with your needs in mind.

  • Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 45 cu. ft.
Capacity (Metric) 1274 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Glass
No. of Doors 2
Shelves 8 Full, 2 Half
Capacity (English) Shelf 100 lb./full shelf; 50 lb./half shelf
Capacity (Metric) Shelf 45 kg/full shelf, 22.5 kg/half shelf
Interior Painted White Steel
Certifications/Compliance cULus, ENERGYSTAR
Color White
Control Type Microprocessor
Ports 1 Rear access port and 2 side access ports
Warranty 2 years parts and labor, plus 3 year compressor warranty
Model CPLA
Access Security Key Lock
Dimensions (D x W x H) Interior 27 x 49 x 61.75 in. (686 x 1245 x 1569 mm)
Dimensions (L x W x H) Exterior 32 x 52.7 x 81 in.(813 x 1339 x 813 mm)
Amperage 5 A
Plug Type NEMA 5-15 P
Electrical Requirements 115 V 60 Hz
Shipping Weight (English) 561 lb.
Shipping Weight (Metric) 255 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.