Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators, Solid-Stainless Steel Door

Catalog No. FBG49RSSA Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
23 cu. ft.
49 cu. ft.
Shelves:
4 Full
8 Full
Color:
Gray
White
Interior:
Painted White Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Shelves Color Interior
FBG49RSSA 49 cu. ft. 8 Full Gray Stainless Steel
FBG25RPSA 23 cu. ft. 4 Full White Painted White Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBG49RSSA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.

  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 49 cu. ft.
Capacity (Metric) 1387 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Solid
No. of Doors 2
Door Material Stainless Steel
Shelves 8 Full
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Interior Stainless Steel
Certifications/Compliance cULus, ENERGYSTAR
Color Gray
Control Type Microprocessor
Ports 1 Rear access port
Warranty 2 years parts and labor, plus 3 year compressor warranty
Model RSSA
Access Security Key Lock
Dimensions (D x W x H) Interior 28.5 x 49 x 61.75 in. (724 x 1245 x 1569 mm)
Dimensions (L x W x H) Exterior 32 x 52.7 x 81 in.(813 x 1339 x 2057 mm)
Amperage 8.8 A
Plug Type NEMA 5-15 P
Electrical Requirements 115 V 60 Hz
Shipping Weight (English) 597 lb.
Shipping Weight (Metric) 271 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.