Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp Value Undercounter Refrigerator PROMO

Catalog No. FBV05RPSS24 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
155.7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
FBV05RPSS24 155.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. FBV05RPSS24

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Chill, freeze and retrieve samples safely, simply and economically with the Fisherbrand™ Isotemp Undercounter Refrigerator.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish
TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose Refrigerator
Capacity (English) 5.5 cu. ft.
Capacity (Metric) 155.7 L
Temperature Range 1°C to 12°C
Temperature (Setpoint) Adjustable
Refrigerant R600a hydrocarbon refrigerant
Door Style Hinged
No. of Doors 1
Hinge Side Right
Shelves 3 Adjustable
Exterior Finish Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Casters 2 Adjustable Feet in Front
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime 10 years
Warranty 12 Months
Defrost Manual
Min. Clearance 4 in. on all sides
Access Security Key Lock
Compressor Hermetic
Dimensions (DxWxH) Exterior 59.69 x 60.02 x 85.1 cm (23.5 x 23.63 x 33.5 in.)
Dimensions (D x W x H) Interior 44.5 x 50.8 x 70.5 cm (17.5 x 20 x 27.75 in.)
Amperage 0.8 A
Plug Type NEMA 5-15 P
Voltage 115 V
Frequency 60 Hz
Energy Usage 0.31 kWh/Day
Shipping Weight (English) 95 lb.
Shipping Weight (Metric) 43 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.