Learn More
Enzo Life Sciences Isotetrandrine (1mg). CAS: 477-57-6
SDP
Supplier:  Enzo Life Sciences BMLG5200001
Encompass_Preferred
Isotetrandrine is a biscoclaurine alkaloid which inhibits G-protein mediated activation of phospholipase A2 (PLA2) but not phospholipase C (PLC) or D (PLD). Cell permeable. Alternative name: Berbamine methyl ether, 6,6',7,12-Tetramethoxy-2,2'-dimethyl-berbaman. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20mg/ml) or ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0887