Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isotetrandrine | 477-57-6 | 5 MG
SDP

Catalog No. 5000421088
Encompass_Preferred

Isotetrandrine is an endogenous metabolite and a bioactive component isolated from the plant Stephania tetrandra. This alkaloid dimer is presented as a white to light yellow solid with a molecular formula of C38H42N2O6 and a molecular weight of 622.75.

  • Bioactive component found in Stephania tetrandra.
  • Categorized as an alkaloid and alkaloid dimer.
  • Molecular formula: C38H42N2O6.
  • Molecular weight: 622.75.
  • Appearance: White to light yellow solid.
  • Purity: ≥97.0%.
  • For research use only.

Catalog No. 50-004-21088 Supplier Medchemexpress LLC Supplier No. HYN60455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isotetrandrine is an endogenous metabolite and a bioactive component isolated from the plant Stephania tetrandra. This alkaloid dimer is presented as a white to light yellow solid with a molecular formula of C38H42N2O6 and a molecular weight of 622.75.

  • Bioactive component found in Stephania tetrandra.
  • Categorized as an alkaloid and alkaloid dimer.
  • Molecular formula: C38H42N2O6.
  • Molecular weight: 622.75.
  • Appearance: White to light yellow solid.
  • Purity: ≥97.0%.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.