Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isotetrandrine | 477-57-6 | MFCD00236404 | 1 MG
SDP

Catalog No. 5000418884
Encompass_Preferred

Isotetrandrine is a bioactive component found in Stephania tetrandra. It is intended for research use only and not for sale to patients.

  • Purity: ≥97.0%
  • Molecular weight: 622.75
  • Formula: C38H42N2O6
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Alkaloids, alkaloid dimers
  • Initial source: Plants, Menispermaceae, Stephania tetrandra S. Moore
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-18884 Supplier Medchemexpress LLC Supplier No. HYN60451MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isotetrandrine is a bioactive component found in Stephania tetrandra. It is intended for research use only and not for sale to patients.

  • Purity: ≥97.0%
  • Molecular weight: 622.75
  • Formula: C38H42N2O6
  • Appearance: Solid
  • Color: White to light yellow
  • Structure classification: Alkaloids, alkaloid dimers
  • Initial source: Plants, Menispermaceae, Stephania tetrandra S. Moore
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.