Learn More
New England Biolabs, Inc. Isothermal Amplification Buffer – 6 ml (4 x 1.5 ml)
SDP
Catalog No. 50591183
Encompass
10X Isothermal Amplification Buffer is optimized for use with Bst 2.0 DNA Polymerase and Bst 2.0 WarmStart DNA Polymerase. This buffer provides superior reaction conditions for isothermal amplication applications.
Catalog No. 50-591-183 Supplier New England Biolabs, Inc. Supplier No. B0537S
10X Isothermal Amplification Buffer is optimized for use with Bst 2.0 DNA Polymerase and Bst 2.0 WarmStart DNA Polymerase. This buffer provides superior reaction conditions for isothermal amplication applications.