Learn More
Dojindo Molecular Technologies Inc Isothiocyanobenzyl-EDTA (10MG), CAS: 105394-74-9, is used for immunoimaging.
SDP
Catalog No. NC1350179
Encompass
Isothiocyanobenzyl-EDTA adds EDTA moieties to proteins through their amine groups. The EDTA-tagged protein can then be labeled with heavy metal ions and radioactive metals, such as 111In and 99Tc, and used for immunoimaging. Dojindo has various reagents for biosensor research. (https://www.dojindo.com/surface-chemistry/)
Catalog No. NC1350179 Supplier Dojindo Molecular Technologies Inc Supplier No. M03010
Isothiocyanobenzyl-EDTA adds EDTA moieties to proteins through their amine groups. The EDTA-tagged protein can then be labeled with heavy metal ions and radioactive metals, such as 111In and 99Tc, and used for immunoimaging. Dojindo has various reagents for biosensor research. (https://www.dojindo.com/surface-chemistry/)