Learn More
Beckman Coulter ISOTON II Electrolyte Diluent, 20 L
Catalog No. 15655119
Change view
Click to view available options
Quantity:
20 L
Catalog No. Quantity
15-655-119 20 L
1 options
Catalog No. 15-655-119 Supplier Beckman Coulter Supplier No. 8546719
A diluent recommended for use with Multisizer 4e and other Coulter Counters. ISOTON II is packaged in a 20 L cubitainer which requires a spigot to dispense.
  • For use with the Multisizer 4e and other Coulter Counters
  • Electrolyte solution for most common applications
  • Packaged in a 20 liter container
  • Spigot recommended for dispensing, sold separately
Specifications
Product Type Electrolyte Diluent
Capacity (Metric) 20 L
Quantity 20 L
For Use With (Equipment) Multisizer™ 4e Coulter Counters