Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isotretinoin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012205 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2205 3 x 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2205 Supplier MilliporeSigma Supelco Supplier No. PHR11883X100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isotretinoin is a vitamin A derivative, obtained from the isomerization of retinoic acid in the body. It is therapeutically used to treat severe recalcitrant nodular acne. It is also reported to exhibit anti-inflammatory and anti-tumor activities.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 4759-48-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C20H28O2
MDL Number MFCD00079542
Melting Point 172°C to 175°C
Synonym 13-cis-Retinoic acid; Isotretinoin
Type Certified Reference Material
Quantity 3 x 100 mg
Formula Weight 300.44
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.