Learn More
Cytek Biosciences Inc Isotype Control In Vivo Ready™ Armenian Hamster IgG Isotype Control (PIP) ….. Cat. No. 40-4888-U500 ….. Size 500 µg
Supplier:  Cytek Biosciences Inc 404888U500
Encompass_Preferred
Isotype Control In Vivo Ready™ Armenian Hamster IgG Isotype Control (PIP) ….. Cat. No. 40-4888-U500 ….. Size 500 µgThe PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.
Catalog No. 50-210-3293