Learn More
Cytek Biosciences Inc Isotype Control PE-Cyanine7 Armenian Hamster IgG Isotype Control (PIP) ..... Cat. No. 60-4888-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 604888U025
Encompass_Preferred
Isotype Control PE-Cyanine7 Armenian Hamster IgG Isotype Control (PIP) ..... Cat. No. 60-4888-U025 ..... Size 25 µgThe PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.
Catalog No. 50-210-3542