Learn More
Bio X Cell Control, Rat IgG1, Anti-Horseradish Peroxidase, 5 MG
Supplier: Bio X Cell BE0088R005MG
InVivoMAb rat IgG1 isotype control, anti-horseradish peroxidase, is provided in a 5 mg size. This antibody solution is designed for use as a control in various immunological assays.
- Antibody host: Rat
- Isotype: IgG1, κ
- Store at 4°C; do not freeze
- Ships with blue ice to maintain 4°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.