Learn More
Bio X Cell Isotype Control, Rat IgG1, Anti-HRP, 100 MG
Supplier: Bio X Cell BE0088R100MG
The HRPN monoclonal antibody reacts with horseradish peroxidase (HRP). This antibody is ideal for use as an isotype-matched control for rat IgG1 antibodies in most in vivo and in vitro applications, as HRP is not expressed by mammals.
- Host species: Rat
- Target: Horseradish peroxidase (HRP)
- Clone: HRPN
- Isotype: Rat IgG1, κ
- Purification: Protein G, purified from cell culture supernatant in an animal-free facility
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin levels: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Conjugation: Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.