Learn More
Bio X Cell Isotype Control, Rat IgG2b, Anti-KLH, 100 MG
Supplier: Bio X Cell BE0090R100MG
This InVivoMAb rat IgG2b isotype control is designed for experiments requiring a control antibody with specificity against keyhole limpet hemocyanin.
- Isotype control: rat IgG2b
- Antigen specificity: anti-keyhole limpet hemocyanin
- Package size: 100 mg
- Antibody host: rat
- Antibody isotype: IgG2b, κ
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.