Learn More
Bio X Cell Isotype Control, Rat IgG2a, Anti-Trinitrophenol, Unconjugated, 95% Purity, 100 MG
Supplier: Bio X Cell BE0089R100MG
This InVivoMAb rat IgG2a isotype control reacts specifically with trinitrophenol, which is not expressed in mammals. It serves as an ideal isotype-matched control for rat IgG2a antibodies in both in vivo and in vitro applications. The product is unconjugated, formulated in PBS at pH 6.5 without stabilizers or preservatives, and features high purity of at least 95% and an endotoxin level of 1 EU/mg.
- Unconjugated.
- Formulated in PBS, pH 6.5, free of stabilizers and preservatives.
- Endotoxin level 1 EU/mg.
- Purity 95% by SDS-PAGE.
- Sterile 0.2 m filtration.
- Animal-free production facility.
- Protein G purification.
- Store at 4C, do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.