Learn More
Bio X Cell Isotype Control, Rat IgG2a, Anti-Trinitrophenol, 50 MG
Supplier: Bio X Cell BE0089R050MG
The 2A3 monoclonal antibody reacts with trinitrophenol. This antibody is suitable as an isotype-matched control for rat IgG2a antibodies in most in vivo and in vitro applications because trinitrophenol is not expressed by mammals.
- Isotype: Rat IgG2a, κ
- Recommended dilution buffer: InVivoPure pH 6.5 Dilution Buffer
- Conjugation: Unconjugated (conjugation services available)
- Formulation: PBS, pH 6.5; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg) (Determined by LAL assay)
- Purity: ≥95% (Determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
- Applications: Flow Cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.