Learn More
Bio X Cell Mouse IgG2a Isotype Control, C1.18.4, 5 MG
Supplier: Bio X Cell BE0085R005MG
The C1.18.4 monoclonal antibody is an ideal non-reactive isotype-matched control for mouse IgG2a antibodies in most in vivo and in vitro applications.
- Ideal for use as a non-reactive isotype-matched control for mouse IgG2a antibodies
- Suitable for most in vivo and in vitro applications
- Purified from cell culture supernatant in an animal-free facility
- Contains no stabilizers or preservatives
- Endotoxin levels ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
- Purity ≥95%, determined by SDS-PAGE
- Protein G purification
- Formulated in PBS, pH 7.0
- Sterility ensured by 0.2 μm filtration
- Molecular weight 150 kDa
- Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.