Learn More
Bio X Cell Isotype Control, Mouse IgG2a, Unconjugated, 0.2 m Filtration, 1.18.4
Supplier: Bio X Cell BE0085R025MG
This high-purity, unconjugated monoclonal antibody serves as a non-reactive isotype-matched control for mouse IgG2a antibodies. It is ideal for most in vivo and in vitro applications, providing a reliable control for preclinical research.
- Clone C1.18.4
- Isotype mouse IgG2a,
- Recommended for use with pH 7.0 dilution buffer
- Unconjugated format
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level of 1EU/mg (0.001EU/g)
- Purity 95%
- Sterile via 0.2 m filtration
- Produced in an animal-free facility
- Purified by protein G
- Molecular weight of 150 kDa
- Store at 4C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.