Learn More
Bio X Cell Isotype Control, MOPC-21, Mouse IgG1, 25 MG
Supplier: Bio X Cell BE0083R025MG
The MOPC-21 monoclonal antibody is an InVivoMAb mouse IgG1 isotype control, designed as a non-reactive isotype-matched control for mouse IgG1 antibodies in most in vivo and in vitro applications.
- Formulated in PBS, pH 6.5, without stabilizers or preservatives
- Unconjugated
- Endotoxin ≤1EU/mg (≤0.001EU/μg)
- Purity ≥95% by SDS-PAGE
- Sterility via 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility using Protein G
- Molecular weight: 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.