Learn More
Bio X Cell Mouse IgG1 Isotype Control, MOPC-21, 50 MG
Supplier: Bio X Cell BE0083R050MG
The MOPC-21 monoclonal antibody is ideal for use as a non-reactive isotype-matched control for mouse IgG1 antibodies in most in vivo and in vitro applications.
- Isotype: mouse IgG1, κ
- Formulation: PBS, pH 6.5, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg), determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 µm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.