Learn More
TARGETMOL CHEMICALS INC Isourolithin A 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1255751MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Isourolithin A is a naturally occurring compound found in a variety of plants, including parsley, celery and chamomile. Isourolithin A is a member of the flavonoid family and is known for its antioxidant and anti-inflammatory properties. Purity 98.25%
Catalog No. 50-287-9856