Learn More
Cayman Chemical IsoursodeoxycholIc AcId 10mg
SDP
Catalog No. 502947776
Encompass_Preferred
An epimer of ursodeoxycholic acid; decreases serum ALP, AST, and ALT levels in bile duct-ligated rats at 2,500 mg/kg in the diet
Catalog No. 50-294-7776 Supplier Cayman Chemical Supplier No. 3462010 MG
An epimer of ursodeoxycholic acid; decreases serum ALP, AST, and ALT levels in bile duct-ligated rats at 2,500 mg/kg in the diet