Learn More
Medchemexpress LLC Isovaleramide (3-Methylbutanamide) | 541-46-8 | 98.0% | 1 ML
SDP
Catalog No. 5000412794
Encompass_Preferred
Isovaleramide (3-Methylbutanamide) is an orally active anticonvulsant that inhibits alcohol dehydrogenase (ADH) activity and regulates the GABAergic system. It reduces acute kidney injury and has antiepileptic, anxiolytic, sedative, and hypnotic effects.
  • Orally active anticonvulsant
  • Inhibits alcohol dehydrogenase (ADH) activity
  • Regulates GABAergic system
  • Reduces acute kidney injury
  • Antiepileptic effects
  • Anxiolytic effects
  • Sedative effects
  • Hypnotic effects
Catalog No. 50-004-12794 Supplier Medchemexpress LLC Supplier No. HYB122910MM/1ML
Add to Cart
Add to Cart
Isovaleramide (3-Methylbutanamide) is an orally active anticonvulsant that inhibits alcohol dehydrogenase (ADH) activity and regulates the GABAergic system. It reduces acute kidney injury and has antiepileptic, anxiolytic, sedative, and hypnotic effects.
  • Orally active anticonvulsant
  • Inhibits alcohol dehydrogenase (ADH) activity
  • Regulates GABAergic system
  • Reduces acute kidney injury
  • Antiepileptic effects
  • Anxiolytic effects
  • Sedative effects
  • Hypnotic effects