Learn More
Medchemexpress LLC Isovaleramide (3-Methylbutanamide) | 541-46-8 | 98.0% | 100 MG
SDP
Catalog No. 5000412922
Encompass_Preferred
Isovaleramide (3-Methylbutanamide) is an orally active anticonvulsant. It inhibits alcohol dehydrogenase (ADH) activity and regulates the GABAergic system. It reduces acute kidney injury and has antiepileptic, anxiolytic, sedative, and hypnotic effects.
  • Orally active anticonvulsant
  • Inhibits alcohol dehydrogenase (ADH) activity
  • Regulates the GABAergic system
  • Reduces acute kidney injury
  • Antiepileptic effects
  • Anxiolytic effects
  • Sedative effects
  • Hypnotic effects
Catalog No. 50-004-12922 Supplier Medchemexpress LLC Supplier No. HYB1229100MG
Add to Cart
Add to Cart
Isovaleramide (3-Methylbutanamide) is an orally active anticonvulsant. It inhibits alcohol dehydrogenase (ADH) activity and regulates the GABAergic system. It reduces acute kidney injury and has antiepileptic, anxiolytic, sedative, and hypnotic effects.
  • Orally active anticonvulsant
  • Inhibits alcohol dehydrogenase (ADH) activity
  • Regulates the GABAergic system
  • Reduces acute kidney injury
  • Antiepileptic effects
  • Anxiolytic effects
  • Sedative effects
  • Hypnotic effects