Learn More
Medchemexpress LLC Isovaleric acid-13C | 87994-84-1 | 99.0% | 103.12 | 10 MG
SDP
Catalog No. 5000451387
Encompass_Preferred
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during the drug development process.
Catalog No. 50-004-51387 Supplier Medchemexpress LLC Supplier No. HYW012980S110MG
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. It is for research use only and not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation during the drug development process.