Learn More
Medchemexpress LLC Isovaleric acid-13C | 87994-84-1 | 99.0% | 103.12 | 100 MG
SDP
Catalog No. 5000454087
Encompass_Preferred
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-004-54087 Supplier Medchemexpress LLC Supplier No. HYW012980S1100MG
Add to Cart
Add to Cart
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.