Learn More
Medchemexpress LLC Isovaleric acid-13C | 87994-84-1 | MFCD00084022 | 99.0% | 103.12 | 50 MG
SDP
Catalog No. 5000367001
Encompass_Preferred
Isovaleric acid-13C is a stable isotope-labeled version of Isovaleric acid, a naturally occurring fatty acid. It is commonly used in research as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. This compound aids in studying metabolic pathways and drug pharmacokinetics.
  • Used as a tracer in various research applications.
  • Functions as an internal standard for quantitative analysis.
  • Beneficial for metabolic studies.
  • Helps in understanding drug pharmacokinetics.
  • Supports analytical methods like NMR, GC-MS, and LC-MS.
Catalog No. 50-003-67001 Supplier Medchemexpress LLC Supplier No. HYW012980S150MG
Isovaleric acid-13C is a stable isotope-labeled version of Isovaleric acid, a naturally occurring fatty acid. It is commonly used in research as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. This compound aids in studying metabolic pathways and drug pharmacokinetics.
  • Used as a tracer in various research applications.
  • Functions as an internal standard for quantitative analysis.
  • Beneficial for metabolic studies.
  • Helps in understanding drug pharmacokinetics.
  • Supports analytical methods like NMR, GC-MS, and LC-MS.