Learn More
Medchemexpress LLC Isovaleric acid | 503-74-2 | ≥98.0% | 102.13 | 500 MG
SDP
Catalog No. 5000367002
Encompass_Preferred
Isovaleric acid is an orally active short-chain fatty acid. It inhibits osteoclast differentiation through AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. This product is suitable for research on skeletal diseases, such as osteoporosis, and various intestinal disorders.
  • Inhibits osteoclast differentiation
  • Stimulates AMPK phosphorylation
  • Promotes colonic smooth muscle relaxation
  • Activates cAMP/PKA pathway
  • Useful in research on skeletal diseases
  • Useful in research on intestinal disorders
Catalog No. 50-003-67002 Supplier Medchemexpress LLC Supplier No. HYW012980R500MG
Isovaleric acid is an orally active short-chain fatty acid. It inhibits osteoclast differentiation through AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. This product is suitable for research on skeletal diseases, such as osteoporosis, and various intestinal disorders.
  • Inhibits osteoclast differentiation
  • Stimulates AMPK phosphorylation
  • Promotes colonic smooth muscle relaxation
  • Activates cAMP/PKA pathway
  • Useful in research on skeletal diseases
  • Useful in research on intestinal disorders