Learn More
Medchemexpress LLC Isovaleric acid | 503-74-2 | ≥98.0% | 102.13 | 500 MG
SDP
Catalog No. 5000451191
Encompass_Preferred
Isovaleric acid is an orally active short-chain fatty acid that inhibits osteoclast differentiation by stimulating AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. It can be used in research on skeletal diseases (such as osteoporosis) and intestinal disorders.
  • Oral active short-chain fatty acid.
  • Inhibits osteoclast differentiation by stimulating AMPK phosphorylation.
  • Promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway.
  • Can be used in research on skeletal diseases (e.g., osteoporosis) and intestinal disorders.
Catalog No. 50-004-51191 Supplier Medchemexpress LLC Supplier No. HYW012980500MG
Add to Cart
Add to Cart
Isovaleric acid is an orally active short-chain fatty acid that inhibits osteoclast differentiation by stimulating AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. It can be used in research on skeletal diseases (such as osteoporosis) and intestinal disorders.
  • Oral active short-chain fatty acid.
  • Inhibits osteoclast differentiation by stimulating AMPK phosphorylation.
  • Promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway.
  • Can be used in research on skeletal diseases (e.g., osteoporosis) and intestinal disorders.