Learn More
Medchemexpress LLC Isovaleric acid | 503-74-2 | 99.8% | 102.13 | 250 MG
SDP
Catalog No. 5000366305
Encompass_Preferred
Isovaleric acid (Standard) is the analytical standard of Isovaleric acid. This product is intended for research and analytical applications. Isovaleric acid is an oral active short-chain fatty acid that inhibits osteoclast differentiation by stimulating AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. It can be used in research on skeletal diseases (such as osteoporosis) and intestinal disorders.
  • Used as a reference standard supplied assay
  • Utilized in qualitative research experiments involving HPLC, GC, and MS
  • Utilized in quantitative research experiments involving HPLC, GC, and MS
  • Utilized in methodological research experiments involving HPLC, GC, and MS
Catalog No. 50-003-66305 Supplier Medchemexpress LLC Supplier No. 250MGHYW012980R
Isovaleric acid (Standard) is the analytical standard of Isovaleric acid. This product is intended for research and analytical applications. Isovaleric acid is an oral active short-chain fatty acid that inhibits osteoclast differentiation by stimulating AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. It can be used in research on skeletal diseases (such as osteoporosis) and intestinal disorders.
  • Used as a reference standard supplied assay
  • Utilized in qualitative research experiments involving HPLC, GC, and MS
  • Utilized in quantitative research experiments involving HPLC, GC, and MS
  • Utilized in methodological research experiments involving HPLC, GC, and MS