Learn More
Medchemexpress LLC Isovaleric acid | 503-74-2 | MFCD00004419 | 99% | 102.13 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012980R50MG
Encompass_Preferred
Isovaleric acid is an oral active short-chain fatty acid that inhibits osteoclast differentiation by stimulating AMPK phosphorylation and promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway. It can be used in research on skeletal diseases (such as osteoporosis) and intestinal disorders. As a laboratory chemical, it is classified as a combustible liquid (Category 4), causing severe skin burns and eye damage (Category 1A, 1B, 1C), and is harmful to aquatic life (Category 3).
  • Oral active short-chain fatty acid
  • Inhibits osteoclast differentiation by stimulating AMPK phosphorylation
  • Promotes colonic smooth muscle relaxation by activating the cAMP/PKA pathway
  • Used in research on skeletal diseases, including osteoporosis
  • Used in research on intestinal disorders
  • Natural fatty acid
  • Analytical standard for research and analytical applications
Catalog No. 50-004-51453