Learn More
Medchemexpress LLC Isovaleric acid-d2 | 95927-02-9 | 99.49% | 104.14 | 100 MG
SDP
Catalog No. 5000454088
Encompass_Preferred
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and known to affect neonatal death and possible Jamaican vomiting sickness in humans. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54088 Supplier Medchemexpress LLC Supplier No. HYW012980S2100MG
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and known to affect neonatal death and possible Jamaican vomiting sickness in humans. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS