Learn More
Medchemexpress LLC Isovaleric acid-d2 | 95927-02-9 | 99.49% | 104.14 | 25 MG
SDP
Catalog No. 5000452711
Encompass_Preferred
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid, a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52711 Supplier Medchemexpress LLC Supplier No. 25MGHYW012980S2
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid, a natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS