Learn More
Medchemexpress LLC Isovaleric acid-d2 | 95927-02-9 | 99.5% | 104.14 | 50 MG
SDP
Catalog No. 5000451192
Encompass_Preferred
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and is known to affect neonatal death and possible Jamaican vomiting sickness in humans. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules largely as tracers for quantitation during the drug development process, and deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable isotope-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration
  • High purity
Catalog No. 50-004-51192 Supplier Medchemexpress LLC Supplier No. HYW012980S250MG
Isovaleric acid-d2 is the deuterium labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and is known to affect neonatal death and possible Jamaican vomiting sickness in humans. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules largely as tracers for quantitation during the drug development process, and deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable isotope-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration
  • High purity