Learn More
Medchemexpress LLC Isovaleric acid-d9 | 344298-81-3 | 98.5% | 111.19 | 100 MG
SDP
Catalog No. 5000451967
Encompass_Preferred
Isovaleric acid-d9 is a deuterium-labeled form of isovaleric acid, a natural fatty acid. This compound is intended for research use only and serves as a valuable tool for quantitative analysis, acting as a tracer or an internal standard.
  • Deuterium labeled.
  • Natural fatty acid derivative.
  • Suitable for use as a tracer in research.
  • Functions as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.
Catalog No. 50-004-51967 Supplier Medchemexpress LLC Supplier No. HYW012980S100MG
Isovaleric acid-d9 is a deuterium-labeled form of isovaleric acid, a natural fatty acid. This compound is intended for research use only and serves as a valuable tool for quantitative analysis, acting as a tracer or an internal standard.
  • Deuterium labeled.
  • Natural fatty acid derivative.
  • Suitable for use as a tracer in research.
  • Functions as an internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS analytical methods.