Learn More
Medchemexpress LLC Isovaleric acid-d9 | 344298-81-3 | 98.5% | 111.19 | 25 MG
SDP
Catalog No. 5000452549
Encompass_Preferred
Isovaleric acid-d9 is a deuterium-labeled version of Isovaleric acid, which is a naturally occurring fatty acid. It is recognized for its potential effects on neonatal death and a condition known as Jamaican vomiting sickness in humans. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled Isovaleric acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52549 Supplier Medchemexpress LLC Supplier No. 25MGHYW012980S
Isovaleric acid-d9 is a deuterium-labeled version of Isovaleric acid, which is a naturally occurring fatty acid. It is recognized for its potential effects on neonatal death and a condition known as Jamaican vomiting sickness in humans. This compound is intended for research use only and is not sold to patients.
  • Deuterium labeled Isovaleric acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.