Learn More
Medchemexpress LLC Isovaleric acid-d9 | 344298-81-3 | 98.5% | 111.19 | 5 MG
SDP
Catalog No. 5000366676
Encompass_Preferred
Isovaleric acid-d9 is a deuterium-labeled natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66676 Supplier Medchemexpress LLC Supplier No. HYW012980S5MG
Isovaleric acid-d9 is a deuterium-labeled natural fatty acid known to affect neonatal death and possible Jamaican vomiting sickness in humans. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.