Learn More
Medchemexpress LLC Isovaleric acid-d9 | 344298-81-3 | 98.5% | 111.19 | 50 MG
SDP
Catalog No. 5000451968
Encompass_Preferred
Isovaleric acid-d9 is a deuterium-labeled version of Isovaleric acid, which is a natural fatty acid known to affect neonatal death and potentially Jamaican vomiting sickness in humans. It is intended for research use only.
  • Deuterium labeled isovaleric acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51968 Supplier Medchemexpress LLC Supplier No. HYW012980S50MG
Isovaleric acid-d9 is a deuterium-labeled version of Isovaleric acid, which is a natural fatty acid known to affect neonatal death and potentially Jamaican vomiting sickness in humans. It is intended for research use only.
  • Deuterium labeled isovaleric acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.