Learn More
Sigma Aldrich Fine Chemicals Biosciences Isovaleryl coenzyme A lith10MG
Catalog No. 502904033
Encompass
Isovaleryl coenzyme A an intermediate of the leucine catabolism. It is synthesized from -methylbutyric acid by the action of enzyme acyl CoA synthetase. The conversion of isovaleryl-CoA to methylcrotonyl-CoA is catalyzed by the enzyme isovaleryl-CoA dehydrogenase in leucine catabolism pathway. The enzyme isovaleryl-CoA oxidase also calalyzes this conversion in fatty acid -oxidation pathway.
Catalog No. 50-290-4033 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I938110MG
Isovaleryl coenzyme A an intermediate of the leucine catabolism. It is synthesized from -methylbutyric acid by the action of enzyme acyl CoA synthetase. The conversion of isovaleryl-CoA to methylcrotonyl-CoA is catalyzed by the enzyme isovaleryl-CoA dehydrogenase in leucine catabolism pathway. The enzyme isovaleryl-CoA oxidase also calalyzes this conversion in fatty acid -oxidation pathway.